BOC-glicina, 99+ %, Thermo Scientific Chemicals
BOC-glicina, 99+ %, Thermo Scientific Chemicals
BOC-glicina, 99+ %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

BOC-glicina, 99+ %, Thermo Scientific Chemicals

CAS: 4530-20-5 | C7H13NO4 | 175.18 g/mol
Have Questions?
Número de catálogoCantidadEnvase
158801000100 gFrasco de vidrio
Número de catálogo 158801000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS4530-20-5
IUPAC Name2-{[(tert-butoxy)carbonyl]amino}acetic acid
Molecular FormulaC7H13NO4
InChI KeyVRPJIFMKZZEXLR-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)NCC(O)=O
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Infrared spectrumConforms
Melting point85°C to 90°C
Titration with NaOH>=99.0 %
HPLC>=99.0 %
Ver más
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only