sym-Diphenylcarbazide, 98%, ACS reagent
sym-Diphenylcarbazide, 98%, ACS reagent
sym-Diphenylcarbazide, 98%, ACS reagent
Thermo Scientific Chemicals

sym-Diphenylcarbazide, 98%, ACS reagent

CAS: 140-22-7 | C13H14N4O | 242.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 158845000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS140-22-7
IUPAC Name1,3-bis(phenylamino)urea
Molecular FormulaC13H14N4O
InChI KeyKSPIHGBHKVISFI-UHFFFAOYSA-N
SMILESO=C(NNC1=CC=CC=C1)NNC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige to pink to
Appearance (Form)Powder or crystals
Infrared spectrumConforms
Melting point173°C to 176°C
HPLC>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only