1,2,3,6-Tetrahidropiridina, 98 %, Thermo Scientific Chemicals
1,2,3,6-Tetrahidropiridina, 98 %, Thermo Scientific Chemicals
1,2,3,6-Tetrahidropiridina, 98 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

1,2,3,6-Tetrahidropiridina, 98 %, Thermo Scientific Chemicals

CAS: 694-05-3 | C5H9N | 83.13 g/mol
Have Questions?
Número de catálogoCantidadEnvase
1588700101 gFrasco de vidrio
Número de catálogo 158870010
Precio (USD)
-
Cantidad:
1 g
Envase:
Frasco de vidrio
Pedido a granel o personalizado
Identificadores químicos
CAS694-05-3
IUPAC Name1,2,3,6-tetrahydropyridine
Molecular FormulaC5H9N
InChI KeyFTAHXMZRJCZXDL-UHFFFAOYSA-N
SMILESC1CC=CCN1
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Color)Clear colorless to yellow to tan
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4790 to 1.4830 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only