m-Xylene, 99%, Extra Dry, AcroSeal™
m-Xylene, 99%, Extra Dry, AcroSeal™
m-Xylene, 99%, Extra Dry, AcroSeal™
Thermo Scientific Chemicals

m-Xylene, 99%, Extra Dry, AcroSeal™

CAS: 108-38-3 | C8H10 | 106.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 158930010
Price (EUR)
-
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS108-38-3
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Water=<0.005 %
GC>=98.5 %
Residue after evaporation=<0.01 %
Appearance (Color)Clear colorless
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only