Catechol, 99+%
Catechol, 99+%
Catechol, 99+%
Catechol, 99+%
Thermo Scientific Chemicals

Catechol, 99+%

CAS: 120-80-9 | C6H6O2 | 110.11 g/mol
数量:
5 g
500 g
2.5 kg
10 kg
パッケージ:
すべて表示
Fibre drum
製品番号(カタログ番号) 158980100
価格(JPY)
-
数量:
10 kg
パッケージ:
Fibre drum
一括またはカスタム形式をリクエストする
化学物質識別子
CAS120-80-9
IUPAC Namebenzene-1,2-diol
Molecular FormulaC6H6O2
InChI KeyYCIMNLLNPGFGHC-UHFFFAOYSA-N
SMILESOC1=CC=CC=C1O
さらに表示
Appearance (Form)Crystalline flakes
Melting point103°C to 106°C
AppearanceMay darken on storage
Infrared spectrumConforms
Appearance (Color)Off-white to grey-brown
さらに表示

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

RUO – Research Use Only