Benzenesulfonic acid, 90%, technical
Benzenesulfonic acid, 90%, technical
Benzenesulfonic acid, 90%, technical
Thermo Scientific Chemicals

Benzenesulfonic acid, 90%, technical

CAS: 98-11-3 | C6H6O3S | 158.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 159021000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS98-11-3
IUPAC Namebenzenesulfonic acid
Molecular FormulaC6H6O3S
InChI KeySRSXLGNVWSONIS-UHFFFAOYSA-N
SMILESOS(=O)(=O)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Low melting solid or fused mass
Titration with CHA>=88 %
Infrared spectrumConforms
Appearance (Color)Off-white to brown
Titration with CHA=<3 % (H2SO4)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only