This product is now part of a food safety range that includes Thermo Scientific™ culture media, molecular solutions, and other products for your complete microbiological workflow. In US, Canada and Europe, order this product from Thermo Fisher Scientific microbiology. >> Order now
Applied Biosystems™

Dynabeads™ MX 8-tube Mixing Wheel

Catalog number:  15904

Related applications:

Food & Beverage

Back to top

Description

Attachment for Dynabeads® MX Mixer Base (Cat. No. 159-02). Designed to hold 8 sample tubes ranging from 15 to 27 mm in diameter. Requires use of Mixer Turret (Cat. No. 159-06) to affix to Mixer Base.
For Research Use Only. Not for use in diagnostic procedures.

Specifications

For Use With (Equipment): Dynabeads® Sample Mixers
Product Size: 1 unit
Shipping Condition: Room Temperature
Type: Sample Mixer Accessories

Contents & storage

Store at room temperature.