4-Chlorobenzoic acid, 99%
4-Chlorobenzoic acid, 99%
4-Chlorobenzoic acid, 99%
Thermo Scientific Chemicals

4-Chlorobenzoic acid, 99%

CAS: 74-11-3 | C7H5ClO2 | 156.57 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
250 g
1 kg
Packaging:
Plastic bottle
Catalog number 159250250
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.50
Save 3.85
Each
Quantity:
25 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (USD)
23.65
Special offer
Online exclusive
Ends: 28-Dec-2026
27.50
Save 3.85
Each
Chemical Identifiers
CAS74-11-3
IUPAC Name4-chlorobenzoic acid
Molecular FormulaC7H5ClO2
InChI KeyXRHGYUZYPHTUJZ-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=C(Cl)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=98.5 %
Melting point240°C to 243°C
Titration with NaOH98.5 to 101.5 %
Water=<0.1 % (K.F.)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only