Hydrindantin dihydrate, 96%
Hydrindantin dihydrate, 96%
Hydrindantin dihydrate, 96%
Thermo Scientific Chemicals

Hydrindantin dihydrate, 96%

CAS: 5950-69-6 | C18H14O8 | 358.29 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 159280500
Price (USD)
150.65
Special offer
Online exclusive
Ends: 30-Jun-2026
177.00
Save 26.35 (15%)
Each
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
150.65
Special offer
Online exclusive
Ends: 30-Jun-2026
177.00
Save 26.35 (15%)
Each
Chemical Identifiers
CAS5950-69-6
IUPAC Name2,2',3,3,3',3'-hexahydroxy-1H,1'H,2H,2'H,3H,3'H-[2,2'-biindene]-1,1'-dione
Molecular FormulaC18H14O8
InChI KeyQHVADKNWNMILPQ-UHFFFAOYNA-N
SMILESOC1(O)C2=CC=CC=C2C(=O)C1(O)C1(O)C(=O)C2=CC=CC=C2C1(O)O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light beige to pink-beige
Infrared spectrumConforms
Loss on drying9 % to 11 % (105°C, 3 hr)
HPLC>=95 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only