2-Amino-3,5-dichloropyridine, 97%
2-Amino-3,5-dichloropyridine, 97%
2-Amino-3,5-dichloropyridine, 97%
Thermo Scientific Chemicals

2-Amino-3,5-dichloropyridine, 97%

CAS: 4214-74-8 | C5H4Cl2N2 | 163.00 g/mol
数量:
25 g
100 g
パッケージ:
すべて表示
Plastic Bottle
製品番号(カタログ番号) 159380250
価格(JPY)
-
数量:
25 g
パッケージ:
Plastic Bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS4214-74-8
IUPAC Name3,5-dichloropyridin-2-amine
Molecular FormulaC5H4Cl2N2
InChI KeyOCWBGKZFOYMCCN-UHFFFAOYSA-N
SMILESNC1=NC=C(Cl)C=C1Cl
さらに表示
Infrared spectrumConforms
Melting point79°C to 83°C
Appearance (Color)Light beige to light brown
HPLC>=96.0 %
Water=<0.5 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only