2-Methyl-5-nitroimidazole, 99%
2-Methyl-5-nitroimidazole, 99%
2-Methyl-5-nitroimidazole, 99%
Thermo Scientific Chemicals

2-Methyl-5-nitroimidazole, 99%

CAS: 88054-22-2 | C4H5N3O2 | 127.10 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
250 g
Packaging:
Plastic bottle
Catalog number 159470500
Price (EUR)
38,20
Each
Quantity:
50 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (EUR)
38,20
Each
Chemical Identifiers
CAS88054-22-2
IUPAC Name2-methyl-5-nitro-1H-imidazole
Molecular FormulaC4H5N3O2
InChI KeyFFYTTYVSDVWNMY-UHFFFAOYSA-N
SMILESCC1=NC=C(N1)[N+]([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige
Infrared spectrumConforms
Titration with HClO4>=98.5 %
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only