alpha-2-Chloroisodurene, 98%
alpha-2-Chloroisodurene, 98%
alpha-2-Chloroisodurene, 98%
Thermo Scientific Chemicals

alpha-2-Chloroisodurene, 98%

CAS: 1585-16-6 | C10H13Cl | 168.66 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 159780250
Price (EUR)
106,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
106,00
Each
Chemical Identifiers
CAS1585-16-6
IUPAC Name2-(chloromethyl)-1,3,5-trimethylbenzene
Molecular FormulaC10H13Cl
InChI KeyUNRGEIXQCZHICP-UHFFFAOYSA-N
SMILESCC1=CC(C)=C(CCl)C(C)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point37°C to 42°C
Appearance (Color)White
Appearance (Form)Fused solid or crystals and/or chunks
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only