Racks and Inserts for Refrigerated and Heated Bath Circulators
Actual product may vary
Racks and Inserts for Refrigerated and Heated Bath Circulators
Racks and Inserts for Refrigerated and Heated Bath Circulators
Thermo Scientific™
Racks and Inserts for Refrigerated and Heated Bath Circulators
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeHoldsFor Use With (Equipment)
1600002RackDepends on insert purchasedA5B,A10B A24B,S49,S19T,S14P,S21P
1600003Rack Insert100-10 mm TubesA5B,A10B A24B,S49,S19T,S14P,S21P
1600004Rack Insert60-16 mm TubesA5B,A10B A24B,S49,S19T,S14P,S21P
1600006Rack InsertDishes, plates, bags, pouchesA5B,A10B A24B,S49,S19T,S14P,S21P
1600079RackDepends on insert purchasedA25B, A40, S21, S30
1600080Rack Insert55-10 mm TubesCold and Heated Bath Circulators
1600081Rack Insert32-16 mm TubesA25B, A40, S21, S30
1600082Rack Insert13-25 mm TubesA25B, A40, S21, S30
1600083Rack InsertDishes, plates, bags, pouchesA25B, A40, S21, S30
1600026RackDepends on insert purchasedS13, S12T, S15
1600084Rack Insert39-10 mm TubesS13, S12T, S15
1600085Rack Insert20-16 mm TubesS13, S12T, S15
1600086Rack Insert8-25 mm TubesS13, S12T, S15
1600087Rack InsertDishes, plates, bags, pouchesS13, S12T, S15
1600066Rack Insert26-4.2 mm TubesGLACIER G50
1600067Rack Insert30-2 mm TubesGLACIER G50
Catalog number 1600002
Price (MXN)
-
Type:
Rack
Holds:
Depends on insert purchased
For Use With (Equipment):
A5B,A10B A24B,S49,S19T,S14P,S21P
These stainless-steel racks and inserts support a range of tubes and are designed for use with Thermo Scientific™ Refrigerated and Heated Bath Circulators.
Specifications
For Use With (Equipment)A5B,A10B A24B,S49,S19T,S14P,S21P
HoldsDepends on insert purchased
MaterialStainless Steel
TypeRack
Unit SizeEach