2-Bromobenzoic acid, 97%
2-Bromobenzoic acid, 97%
2-Bromobenzoic acid, 97%
Thermo Scientific Chemicals

2-Bromobenzoic acid, 97%

CAS: 88-65-3 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Packaging:
View all
Glass bottle
Catalog number 160080250
Price (EUR)
29,65
Online exclusive
32,80
Save 3,15
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
29,65
Online exclusive
32,80
Save 3,15
Each
Chemical Identifiers
CAS88-65-3
IUPAC Name2-bromobenzoic acid
Molecular FormulaC7H5BrO2
InChI KeyXRXMNWGCKISMOH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1Br
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to yellow to beige
Melting point145°C to 150°C
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only