Ácido de 2-bromobenzoico, 97 %, Thermo Scientific Chemicals
Ácido de 2-bromobenzoico, 97 %, Thermo Scientific Chemicals
Ácido de 2-bromobenzoico, 97 %, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Ácido de 2-bromobenzoico, 97 %, Thermo Scientific Chemicals

CAS: 88-65-3 | C7H5BrO2 | 201.02 g/mol
Have Questions?
Cambiar vistabuttonViewtableView
Cantidad:
25 g
100 g
Envase:
Ver todo
Frasco de plástico
Número de catálogo 160081000
Precio (USD)
-
Cantidad:
100 g
Envase:
Frasco de plástico
Pedido a granel o personalizado
Identificadores químicos
CAS88-65-3
IUPAC Name2-bromobenzoic acid
Molecular FormulaC7H5BrO2
InChI KeyXRXMNWGCKISMOH-UHFFFAOYSA-N
SMILESOC(=O)C1=CC=CC=C1Br
Ver más
EspecificacionesSpecification SheetHoja de especificaciones
Appearance (Form)Powder
Infrared spectrumConforms
HPLC>=96.0 %
Appearance (Color)White to yellow to beige
Melting point145°C to 150°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only