Newborn Calf Serum, New Zealand origin, 500 mL - FAQs
Newborn Calf Serum, New Zealand origin, 500 mL - FAQs
View additional product information for Newborn Calf Serum, New Zealand origin - FAQs (16010159, 16010167, 16010142)