Methyl vanillate, 99%
Methyl vanillate, 99%
Methyl vanillate, 99%
Thermo Scientific Chemicals

Methyl vanillate, 99%

CAS: 3943-74-6 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 160300050
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.70
Save 6.05
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
32.65
Special offer
Online exclusive
Ends: 28-Dec-2026
38.70
Save 6.05
Each
Chemical Identifiers
CAS3943-74-6
IUPAC Namemethyl 4-hydroxy-3-methoxybenzoate
Molecular FormulaC9H10O4
InChI KeyBVWTXUYLKBHMOX-UHFFFAOYSA-N
SMILESCOC(=O)C1=CC=C(O)C(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder and/or granules
Melting point64°C to 67°C
Appearance (Color)Light yellow to beige
Infrared spectrumConforms
GC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only