1,3,5-Trimethoxybenzene, 99%
1,3,5-Trimethoxybenzene, 99%
1,3,5-Trimethoxybenzene, 99%
Thermo Scientific Chemicals

1,3,5-Trimethoxybenzene, 99%

CAS: 621-23-8 | C9H12O3 | 168.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
Glass bottle
Catalog number 160550500
Price (USD)
-
Quantity:
50 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS621-23-8
IUPAC Name1,3,5-trimethoxybenzene
Molecular FormulaC9H12O3
InChI KeyLKUDPHPHKOZXCD-UHFFFAOYSA-N
SMILESCOC1=CC(OC)=CC(OC)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream to yellow
GC>=98.5 %
Appearance (Form)Powder or crystals or flakes and/or chunks
Infrared spectrumConforms
Melting point50°C to 54°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only