Isobutyryl chloride, 98%
Isobutyryl chloride, 98%
Isobutyryl chloride, 98%
Thermo Scientific Chemicals

Isobutyryl chloride, 98%

CAS: 79-30-1 | C4H7ClO | 106.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 160660100
Price (USD)
28.65
Online exclusive
31.20
Save 2.55
Each
Quantity:
10 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
28.65
Online exclusive
31.20
Save 2.55
Each
Chemical Identifiers
CAS79-30-1
IUPAC Name2-methylpropanoyl chloride
Molecular FormulaC4H7ClO
InChI KeyDGMOBVGABMBZSB-UHFFFAOYSA-N
SMILESCC(C)C(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Color scale=<50 APHA
Infrared spectrumConforms
GC>=97.5 %
Refractive index1.4060 to 1.4080 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only