3-Indolebutyric acid, 98%
3-Indolebutyric acid, 98%
3-Indolebutyric acid, 98%
Thermo Scientific Chemicals

3-Indolebutyric acid, 98%

CAS: 133-32-4 | C12H13NO2 | 203.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 160690050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS133-32-4
IUPAC Name4-(1H-indol-3-yl)butanoic acid
Molecular FormulaC12H13NO2
InChI KeyJTEDVYBZBROSJT-UHFFFAOYSA-N
SMILESOC(=O)CCCC1=CNC2=CC=CC=C12
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point120°C to 125°C
Appearance (Color)White to yellow to beige to brown
Appearance (Form)Powder
Infrared spectrumConforms
Titration with NaOH>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only