Tetraethylammonium bromide, 98%
Tetraethylammonium bromide, 98%
Tetraethylammonium bromide, 98%
Thermo Scientific Chemicals

Tetraethylammonium bromide, 98%

Phase transfer catalyst | CAS: 71-91-0 | C8H20BrN | 210.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
5 kg
Packaging:
View all
Plastic bottle
Catalog number 161072500
Price (TWD)
-
Quantity:
250 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS71-91-0
IUPAC Nametetraethylazanium bromide
Molecular FormulaC8H20BrN
InChI KeyHWCKGOZZJDHMNC-UHFFFAOYSA-M
SMILES[Br-].CC[N+](CC)(CC)CC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder or crystals or solid
Loss on drying=<2 %
Titration Argentometric>=97.5 %
Infrared spectrumConforms
Appearance (Color)White to cream
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only