Veratrole, 99+%
Veratrole, 99+%
Veratrole, 99+%
Thermo Scientific Chemicals

Veratrole, 99+%

CAS: 91-16-7 | C8H10O2 | 138.17 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
1 kg
Packaging:
Glass bottle
Catalog number 161111000
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.90
Save 5.25
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
31.65
Special offer
Online exclusive
Ends: 28-Dec-2026
36.90
Save 5.25
Each
Chemical Identifiers
CAS91-16-7
IUPAC Name1,2-dimethoxybenzene
Molecular FormulaC8H10O2
InChI KeyABDKAPXRBAPSQN-UHFFFAOYSA-N
SMILESCOC1=CC=CC=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid or solid
Infrared spectrumConforms
GC>=99.0 %
Color scale=<50 APHA
Refractive index1.5320 to 1.5360 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only