1-Phenylcyclopentanecarboxylic acid, 98%
1-Phenylcyclopentanecarboxylic acid, 98%
1-Phenylcyclopentanecarboxylic acid, 98%
Thermo Scientific Chemicals

1-Phenylcyclopentanecarboxylic acid, 98%

CAS: 77-55-4 | C12H14O2 | 190.24 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 161140250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS77-55-4
IUPAC Name1-phenylcyclopentane-1-carboxylic acid
Molecular FormulaC12H14O2
InChI KeyRHPCYZLXNNRRMB-UHFFFAOYSA-N
SMILESOC(=O)C1(CCCC1)C1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with NaOH>=97.5 %
Appearance (Form)Powder
Melting point157°C to 161°C
Appearance (Color)Cream to grey-brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only