Tetrabutylammonium iodide, 98%
Tetrabutylammonium iodide, 98%
Tetrabutylammonium iodide, 98%
Thermo Scientific Chemicals

Tetrabutylammonium iodide, 98%

CAS: 311-28-4 | C16H36IN | 369.38 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 161271000
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.60
Save 13.95
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
79.65
Special offer
Online exclusive
Ends: 28-Dec-2026
93.60
Save 13.95
Each
Chemical Identifiers
CAS311-28-4
IUPAC Nametetrabutylazanium iodide
Molecular FormulaC16H36IN
InChI KeyDPKBAXPHAYBPRL-UHFFFAOYSA-M
SMILES[I-].CCCC[N+](CCCC)(CCCC)CCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Powder or crystals or flakes
Infrared spectrumConforms
Melting point141°C to 150°C
Titration Argentometric>=97.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only