3-Phenyl-1-propanol, 98%
3-Phenyl-1-propanol, 98%
3-Phenyl-1-propanol, 98%
Thermo Scientific Chemicals

3-Phenyl-1-propanol, 98%

CAS: 122-97-4 | C9H12O | 136.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
1 kg
Packaging:
Glass bottle
Catalog number 161310010
Price (EUR)
108,00
Each
Quantity:
1 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
108,00
Each
Chemical Identifiers
CAS122-97-4
IUPAC Name3-phenylpropan-1-ol
Molecular FormulaC9H12O
InChI KeyVAJVDSVGBWFCLW-UHFFFAOYSA-N
SMILESOCCCC1=CC=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless
Refractive index1.5250 to 1.5270 (20°C, 589 nm)
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only