4-Methoxystyrene, 96%, stabilized
4-Methoxystyrene, 96%, stabilized
4-Methoxystyrene, 96%, stabilized
Thermo Scientific Chemicals

4-Methoxystyrene, 96%, stabilized

CAS: 637-69-4 | C9H10O | 134.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 161390250
Price (EUR)
258,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
258,00
Each
Chemical Identifiers
CAS637-69-4
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
GC>=95 %
Appearance (Color)Clear colorless to yellow
Infrared spectrumConforms
Refractive index1.5600 to 1.5620 (20°C, 589 nm)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only