Ponceau S, pure
Ponceau S, pure
Ponceau S, pure
Thermo Scientific Chemicals

Ponceau S, pure

CAS: 6226-79-5 | C22H12N4Na4O13S4 | 760.552 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
25 g
100 g
Catalog number 161470250
Price (USD)
52.40
Each
Quantity:
25 g
Request bulk or custom format
Price (USD)
52.40
Each
Chemical Identifiers
CAS6226-79-5
IUPAC Nametetrasodium (4Z)-3-oxo-4-(2-{2-sulfonato-5-[2-(4-sulfonatophenyl)diazen-1-yl]phenyl}hydrazin-1-ylidene)-3,4-dihydronaphthalene-2,7-disulfonate
Molecular FormulaC22H12N4Na4O13S4
InChI KeyLZYGEJPERQQCIY-STNZDNLRSA-J
SMILES[Na+].[Na+].[Na+].[Na+].[O-]S(=O)(=O)C1=CC=C(C=C1)N=NC1=CC=C(C(N\N=C2/C(=O)C(=CC3=CC(=CC=C23)S([O-])(=O)=O)S([O-])(=O)=O)=C1)S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
UV spectrumComplies
Dye content>=70 %
Appearance (Color)Bordeaux to dark brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only