N-(2-Aminoethyl)piperidine, 98%
N-(2-Aminoethyl)piperidine, 98%
N-(2-Aminoethyl)piperidine, 98%
Thermo Scientific Chemicals

N-(2-Aminoethyl)piperidine, 98%

CAS: 27578-60-5 | C7H16N2 | 128.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
25 mL
Packaging:
Glass bottle
Catalog number 161610250
Price (USD)
-
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS27578-60-5
IUPAC Name2-(piperidin-1-yl)ethan-1-amine
Molecular FormulaC7H16N2
InChI KeyCJNRGSHEMCMUOE-UHFFFAOYSA-N
SMILESNCCN1CCCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4730 to 1.4750 (20°C, 589 nm)
Appearance (Color)Clear colorless to yellow
Appearance (Form)Liquid
Infrared spectrumConforms
Titration with HClO4>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only