Atrazine, 95+%
Thermo Scientific Chemicals

Atrazine, 95+%

CAS: 1912-24-9 | C8H14ClN5 | 215.69 g/mol
Have Questions?
Catalog NumberQuantity
1616800505 g
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 161680050
Price (USD)
-
Quantity:
5 g
Request bulk or custom format
Chemical Identifiers
CAS1912-24-9
IUPAC Name6-chloro-N2-ethyl-N4-(propan-2-yl)-1,3,5-triazine-2,4-diamine
Molecular FormulaC8H14ClN5
InChI KeyMXWJVTOOROXGIU-UHFFFAOYSA-N
SMILESCCNC1=NC(Cl)=NC(NC(C)C)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals
Infrared spectrumConforms
HPLC>=95.0 %
Water=<1 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only