Thermo Scientific™

Atrazine, 95+%, Thermo Scientific™

25 g, Each
Thermo Scientific™

Atrazine, 95+%, Thermo Scientific™

25 g, Each
Quantity
Unit Size

Chemical Identifiers

CAS
1912-24-9
Appearance (Color)
White to off-white
Melting point
173°C to 180°C
Infrared spectrum
Conforms
HPLC
>=95.0 %
Appearance (Form)
Powder or crystals

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
RUO – Research Use Only

Figures