Cytosine, 99+%
Cytosine, 99+%
Cytosine, 99+%
Thermo Scientific Chemicals

Cytosine, 99+%

CAS: 71-30-7 | C4H5N3O | 111.10 g/mol
数量:
5 g
25 g
100 g
パッケージ:
Glass bottle
製品番号(カタログ番号) 161760250
価格(JPY)
-
数量:
25 g
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS71-30-7
IUPAC Name6-amino-1,2-dihydropyrimidin-2-one
Molecular FormulaC4H5N3O
InChI KeyOPTASPLRGRRNAP-UHFFFAOYSA-N
SMILESNC1=CC=NC(=O)N1
さらに表示
Loss on drying=<0.5 % (105°C, 5 h)
Appearance (Color)White to light yellow
Appearance (Form)Powder or crystalline powder
Infrared spectrumConforms
HPLC>=99.0 %
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only