Chlorosulfonyl isocyanate, 98+%
Chlorosulfonyl isocyanate, 98+%
Chlorosulfonyl isocyanate, 98+%
Thermo Scientific Chemicals

Chlorosulfonyl isocyanate, 98+%

CAS: 1189-71-5 | CClNO3S | 141.53 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 mL
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 162040010
Price (USD)
1,110.00
Each
Quantity:
1 L
Packaging:
Glass bottle
Request bulk or custom format
Price (USD)
1,110.00
Each
Chemical Identifiers
CAS1189-71-5
IUPAC Name[(chlorosulfonyl)imino]methanone
Molecular FormulaCClNO3S
InChI KeyWRJWRGBVPUUDLA-UHFFFAOYSA-N
SMILESClS(=O)(=O)N=C=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration with NaOH>=98.0 %
Refractive index1.4450 to 1.4490 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Alkylation reagent, C-X bond formation reagent
RUO – Research Use Only