1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
Acros Organics

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3 | CAS: 5108-96-3 | C5H12N2S2 | 164.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
500 g
Packaging:
Glass bottle
Catalog number 162051000
HSN 29339900
Price (INR)
28,760.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
28,760.00
Each
Chemical Identifiers
CAS5108-96-3
IUPAC Namepyrrolidine-1-carbodithioic acid amine
Molecular FormulaC5H12N2S2
InChI KeyMDDIUTVUBYEEEM-UHFFFAOYSA-N
SMILESN.SC(=S)N1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to off-white
Infrared spectrumConforms
Appearance (Form)Crystalline powder
Titration with HClO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only