1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%
Thermo Scientific Chemicals

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%

1-Pyrrolidinecarbodithioic acid, ammonium salt, 98%, C5H12N2S2, CAS Number-5108-96-3 | CAS: 5108-96-3 | C5H12N2S2 | 164.28 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
Packaging:
Glass bottle
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 162055000
Price (USD)
-
Quantity:
500 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS5108-96-3
IUPAC Namepyrrolidine-1-carbodithioic acid amine
Molecular FormulaC5H12N2S2
InChI KeyMDDIUTVUBYEEEM-UHFFFAOYSA-N
SMILESN.SC(=S)N1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Appearance (Color)White to off-white
Infrared spectrumConforms
Titration with HClO497.5 to 102.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only