Goat Serum, New Zealand origin
Gibco™

Goat Serum, New Zealand origin

Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers withRead more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
16210072500 mL
16210064100 mL
Catalog number 16210072
Price (USD)
281.65
Online exclusive
296.00
Save 14.35
Each
Quantity:
500 mL
Price (USD)
281.65
Online exclusive
296.00
Save 14.35
Each
Gibco™ sera offers excellent value for basic cell culture, specialty research, and specific assays, earning the trust of researchers with consistent quality and award-winning support that helps meet your research needs and budget requirements

Sera Category: Specialty

Origin: New Zealand.

For research use or further manufacturing use only. Serum and blood proteins are not for direct administration into humans or animals.
Specifications
Product LineGibco
Shipping ConditionFrozen
SpeciesGoat/Caprine
AgeAdult
Country of OriginNew Zealand
FormLiquid
Product TypeGoat Serum
Quantity500 mL
Serum TreatmentStandard (Sterile-filtered)
SterilitySterile
Unit SizeEach
Contents & Storage
Storage conditions: ≤-10°C
Shipping conditions: Frozen

Frequently asked questions (FAQs)

Does Goat Serum, New Zealand origin (Cat. Nos. 16210064, 16210072) need to be heat-inactivated before use, or is it ready for use as is? If it does need to be heat-inactivated, what is the protocol to do so?

Goat Serum, New Zealand origin is not heat-inactivated. You can heat-inactivate by incubating the fully thawed, unopened serum bottle in a 56 degrees C water bath for 30 min. Be sure to swirl the bottle several times during the 30 min incubation.

Find additional tips, troubleshooting help, and resources within our Cell Culture Support Center.

Will improper storage of my tissue culture reagents affect the growth rate of my culture?

Yes. Store animal sera at -5 to -20 degrees C. Store media at 2 to 8 degrees C; use within recommended shelf life period. Store complete media (supplemented) at 2 to 8 degrees C, and for complete medium the recommended shelf life is 2 to 4 weeks. Additionally, minimize exposure of sera and media to light.