2,2'-Dithiodipyridine, 98%
2,2'-Dithiodipyridine, 98%
2,2'-Dithiodipyridine, 98%
Thermo Scientific Chemicals

2,2'-Dithiodipyridine, 98%

Have Questions?
變更視圖buttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
產品號碼 162230250
價格 (TWD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
化學識別
CAS2127-03-9
IUPAC Name2-(pyridin-2-yldisulfanyl)pyridine
Molecular FormulaC10H8N2S2
InChI KeyHAXFWIACAGNFHA-UHFFFAOYSA-N
SMILESS(SC1=CC=CC=N1)C1=CC=CC=N1
檢視更多
規格Specification Sheet規格表
Melting point55°C to 60°C
Infrared spectrumConforms
GC>=97.5 %
Appearance (Color)White to yellow to beige
Appearance (Form)Powder
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only