1620A Digital Thermometer-Hygrometer
1620A Digital Thermometer-Hygrometer
Fluke™

1620A Digital Thermometer-Hygrometer

The most accurate temperature and humidity graphical data loggers on the market.
Have Questions?
Cambiar vistabuttonViewtableView
Número de catálogoTipoExactitud de temperatura
1622A-H-256Kit de alta precisión Value, inalámbrico±0,25 °C de 15 °C a 35 °C
1620A-H-256Alta Precisión±0,125 °C de 16 °C a 24 °C
Número de catálogo 1622A-H-256
Precio (USD)
-
Tipo:
Kit de alta precisión Value, inalámbrico
Exactitud de temperatura:
±0,25 °C de 15 °C a 35 °C
Thermo hygrometer features superior accuracy, network enabled, powerful logging and analysis tools, two interchangeable calibrated sensors, huge memory, and upgraded software.

The most accurate temperature and humidity graphical data loggers on the market

The Fluke 1620A (DewK) thermo hygrometer and humidity monitor measures temperature to ± 0.125 °C and humidity to ± 1.5 % on two channels, and displays it on a big, clear screen. Ethernet, wireless (optional), or RS-232 will allow you to network as many Fluke 1620s (DewKs) as you’d like. And the LogWare III software will allow real-time logging of multiple Fluke 1620s (DewKs), data storage to your computer and seamless integration with MET/CAL™ software.

Thermo hygrometer features at a glance:
• Superior accuracy
• Network enabled
• Powerful logging and analysis tools
• Two interchangeable calibrated sensors
• Huge memory
• Upgraded software

Two types of sensors are available.
The high-accuracy sensor (H model) reads temperature to ± 0.125 °C over a calibrated range of 16 °C to 24 °C. Relative humidity readings are to ± 1.5 % RH from 20 % RH to 70 % RH.

The standard-accuracy sensor (“S model) reads temperature to ± 0.25 °C over its calibrated range of 15 °C to 35 °C. Relative humidity readings are to ± 2 % RH from 20 % RH to 70 % RH.

All Fluke 1620A (DewK) thermo-hygrometer sensors come with NVLAP accredited certificates of calibration for both temperature and humidity, complete with data and NIST traceability.

Ethernet and wireless capability
With its built-in Ethernet RJ45 jack, multiple Fluke 1620A (DewK) thermo hygrometers can be monitored from the same screen using LogWare III client-server software. Ethernet also gives you the possibility for remote connectivity over the internet, so you can monitor critical conditions while you’re away.

Cables running along the floor can be a safety hazard, and cables hanging from the ceiling and walls are an eyesore. With the Fluke 1620 (DewK), simply connect your computer through an RF modem up to 100 ft away, without the clutter of all the extra cables.

If you need a printout, send data to a printer through the RS-232 interface in real-time.

Math and statistical functions
In addition to temperature and humidity, the DewK calculates dew point, heat index, and rates of change for both temperature and humidity. Min, max, and a variety of other statistics are also calculated and can be shown on-screen. Daily summary statistics, including min, max, and maximum rates of change are stored for the most recent sixty days.

Calibrated sensors
With inputs for two sensors, each measuring both temperature and relative humidity, one Fluke 1620 (DewK)can monitor two locations at the same time. Both sensors can be run via extension cables to remote locations up to 100 feet away, or one sensor can be directly mounted to the top of the DewK.

Each sensor is calibrated for both temperature and humidity by Fluke Calibration. The calibration constants assigned to the sensors reside in a memory chip located inside the sensor housing, so sensors may be used interchangeably between different Fluke 1620 (DewKs), and the recalibration of sensors doesn’t require an accompanying Fluke 1620 (DewK).

Sensors may also be assigned a unique identifier (up to 16 characters) to facilitate record keeping by matching the sensor identifier with the collected data. Each DewK ships with one sensor, with additional sensors available from Fluke Calibration. Spare sensors may also be purchased as a kit, which includes a wall mounting bracket and a 25-foot extension cable.

The DewK thermo hygrometer and humidity monitor has enough on-board memory to store up to 400,000 date- and times stamped data points. That’s two years’ worth of data for both measurements from two sensors if readings are taken every ten minutes!

Alarms and battery backup
Alarm settings can be set up quickly in the Fluke 1620 (DewK)based on temperature, the rate of change in temperature, RH, the rate of change in RH, and instrument fault conditions. Alarms can be both visual (flashing display) and audible (beeping). Likewise, alarm settings can be set up and events triggered in LogWare III. The Fluke 1620 (DewK)is also equipped with a 0 to 12 volt alarm output that can trigger a process control system

A backup battery shuts down the Fluke 1620 (DewKs)display but maintains measurements for up to 16 hours in the event of a power failure.

Especificaciones
Tipos de alarmaPassword-protected visual, audible, and external alarms for temperature, temperature rate, RH, RH rate, and fault conditions
Batería, tipoStandard 9V battery enables continued measuring during power disruptions
Certificaciones/conformidadCertificate of NIST-traceable NVLAP accredited temperature and humidity calibration included
Descripción1622A-H DewK High Accuracy Value Kit, wireless option and USB wireless modem
Tipo de pantalla240 x 128 Graphics Monochrome LCD
Incluye1620A-H Thermo-Hygrometer, High-Accuracy, 2627-S Spare Sensor Kit, 9936A LogWare III Software, Single License, USB to Wireless and RS232 to Wireless Modems
Resolución0.01
Exactitud de temperatura±0,25 °C de 15 °C a 35 °C
Intervalo de humedad0 to 100%
Intervalo de temperatura (métrico)0°C to 50°C
TipoKit de alta precisión Value, inalámbrico
Unit SizeEach