Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech.
Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech.
Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech.
Thermo Scientific Chemicals

Ethyl N-benzyl-3-oxo-4-piperidinecarboxylate hydrochloride, 97%, tech.

CAS: 52763-21-0 | C15H20ClNO3 | 297.78 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 162301000
Price (TWD)
-
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS52763-21-0
IUPAC Namehydrogen ethyl 1-benzyl-3-oxopiperidine-4-carboxylate chloride
Molecular FormulaC15H20ClNO3
InChI KeyUQOMEAWPKSISII-UHFFFAOYNA-N
SMILES[H+].[Cl-].CCOC(=O)C1CCN(CC2=CC=CC=C2)CC1=O
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO4>=96.0 %
Appearance (Form)Crystalline powder
Melting point160°C to 165°C
Appearance (Color)White to brown
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only