N,N'-Diaminoguanidine monohydrochloride, 98%
N,N'-Diaminoguanidine monohydrochloride, 98%
N,N'-Diaminoguanidine monohydrochloride, 98%
Thermo Scientific Chemicals

N,N'-Diaminoguanidine monohydrochloride, 98%

CAS: 36062-19-8 | CH7N5·HCl | 125.56 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 162400250
Price (USD)
-
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS36062-19-8
IUPAC Namehydrogen (E)-N,N''-diaminoguanidine chloride
Molecular FormulaCH8ClN5
InChI KeyHAZRIBSLCUYMQP-UHFFFAOYSA-N
SMILES[H+].[Cl-].NN\C(N)=N\N
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Titration Argentometric97.5 to 102.5 %
Appearance (Color)White to almost white
Melting point178°C to 182°C
Appearance (Form)Powder and/or chunks
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only