Trimethylacetic anhydride, 99%
Trimethylacetic anhydride, 99%
Trimethylacetic anhydride, 99%
Thermo Scientific Chemicals

Trimethylacetic anhydride, 99%

CAS: 1538-75-6 | C10H18O3 | 186.25 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 mL
250 mL
1 L
Packaging:
Glass bottle
Catalog number 162442500
Price (USD)
-
Quantity:
250 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS1538-75-6
IUPAC Name2,2-dimethylpropanoyl 2,2-dimethylpropanoate
Molecular FormulaC10H18O3
InChI KeyPGZVFRAEAAXREB-UHFFFAOYSA-N
SMILESCC(C)(C)C(=O)OC(=O)C(C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.4080 to 1.4100 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only