Hexamethylphosphorous triamide, 97%
Hexamethylphosphorous triamide, 97%
Hexamethylphosphorous triamide, 97%
Thermo Scientific Chemicals

Hexamethylphosphorous triamide, 97%

CAS: 1608-26-0 | C6H18N3P | 163.21 g/mol
数量:
10 mL
50 mL
100 mL
パッケージ:
Glass bottle
製品番号(カタログ番号) 162480500
価格(JPY)
-
数量:
50 mL
パッケージ:
Glass bottle
一括またはカスタム形式をリクエストする
化学物質識別子
CAS1608-26-0
IUPAC Name[bis(dimethylamino)phosphanyl]dimethylamine
Molecular FormulaC6H18N3P
InChI KeyXVDBWWRIXBMVJV-UHFFFAOYSA-N
SMILESCN(C)P(N(C)C)N(C)C
さらに表示
Appearance (Color)Clear colorless to light yellow
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
さらに表示
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only