Hexamethylphosphorous triamide, 97%
Hexamethylphosphorous triamide, 97%
Hexamethylphosphorous triamide, 97%
Thermo Scientific Chemicals

Hexamethylphosphorous triamide, 97%

CAS: 1608-26-0 | C6H18N3P | 163.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 mL
50 mL
100 mL
Packaging:
Glass bottle
Catalog number 162481000
Price (EUR)
326,00
Each
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
326,00
Each
Chemical Identifiers
CAS1608-26-0
IUPAC Name[bis(dimethylamino)phosphanyl]dimethylamine
Molecular FormulaC6H18N3P
InChI KeyXVDBWWRIXBMVJV-UHFFFAOYSA-N
SMILESCN(C)P(N(C)C)N(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
AppearanceMay develop some turbidity or precipitate
Appearance (Form)Liquid
GC>=96.0 %
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only