Homovanillic acid, 98%
Homovanillic acid, 98%
Homovanillic acid, 98%
Thermo Scientific Chemicals

Homovanillic acid, 98%

CAS: 306-08-1 | C9H10O4 | 182.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 162530010
Price (USD)
-
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS306-08-1
IUPAC Name2-(4-hydroxy-3-methoxyphenyl)acetic acid
Molecular FormulaC9H10O4
InChI KeyQRMZSPFSDQBLIX-UHFFFAOYSA-N
SMILESCOC1=CC(CC(O)=O)=CC=C1O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
HPLC>=97.5 %
Appearance (Color)White to beige
Infrared spectrumConforms
Melting point141°C to 145°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only