3,4-Dimethoxybenzaldehyde, 99+%
3,4-Dimethoxybenzaldehyde, 99+%
3,4-Dimethoxybenzaldehyde, 99+%
3,4-Dimethoxybenzaldehyde, 99+%
Thermo Scientific Chemicals

3,4-Dimethoxybenzaldehyde, 99+%

CAS: 120-14-9 | C9H10O3 | 166.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
Packaging:
View all
Plastic bottle
Catalog number 162615000
Price (USD)
-
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Chemical Identifiers
CAS120-14-9
IUPAC Name3,4-dimethoxybenzaldehyde
Molecular FormulaC9H10O3
InChI KeyWJUFSDZVCOTFON-UHFFFAOYSA-N
SMILESCOC1=CC=C(C=O)C=C1OC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point42°C to 45°C
GC>=99.0 %
Appearance (Color)White to yellow to light orange to tan
Appearance (Form)Powder or crystals or flakes and/or lumps
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only