2,4-Dichloropyrimidine, 97%
2,4-Dichloropyrimidine, 97%
2,4-Dichloropyrimidine, 97%
Thermo Scientific Chemicals

2,4-Dichloropyrimidine, 97%

CAS: 3934-20-1 | C4H2Cl2N2 | 148.98 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 162651000
Price (EUR)
392,00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
392,00
Each
Chemical Identifiers
CAS3934-20-1
IUPAC Name2,4-dichloropyrimidine
Molecular FormulaC4H2Cl2N2
InChI KeyBTTNYQZNBZNDOR-UHFFFAOYSA-N
SMILESClC1=CC=NC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
GC>=96.0 %
Appearance (Color)White to yellow to beige to grey
Melting point57°C to 62°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only