Propargyl chloride, 70 wt.% solution in toluene
Propargyl chloride, 70 wt.% solution in toluene
Propargyl chloride, 70 wt.% solution in toluene
Thermo Scientific Chemicals

Propargyl chloride, 70 wt.% solution in toluene

CAS: 624-65-7 | C3H3Cl | 74.51 g/mol
Catalog NumberQuantityPackaging
162701000100 mLGlass bottle
Catalog number 162701000
Price (JPY)
-
Quantity:
100 mL
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS624-65-7
IUPAC Name3-chloroprop-1-yne
Molecular FormulaC3H3Cl
InChI KeyLJZPPWWHKPGCHS-UHFFFAOYSA-N
SMILESClCC#C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Assay68 to 72 %
Appearance (Color)Clear colorless to light yellow
Refractive index1.4550 to 1.4570 (20°C, 589 nm)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only