4,6-Dichloro-2-(methylthio)pyrimidine, 98%
4,6-Dichloro-2-(methylthio)pyrimidine, 98%
4,6-Dichloro-2-(methylthio)pyrimidine, 98%
Thermo Scientific Chemicals

4,6-Dichloro-2-(methylthio)pyrimidine, 98%

CAS: 6299-25-8 | C5H4Cl2N2S | 195.07 g/mol
Have Questions?
Catalog NumberQuantityPackaging
1630300505 gGlass bottle
Catalog number 163030050
Price (USD)
-
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS6299-25-8
IUPAC Name4,6-dichloro-2-(methylsulfanyl)pyrimidine
Molecular FormulaC5H4Cl2N2S
InChI KeyFCMLONIWOAGZJX-UHFFFAOYSA-N
SMILESCSC1=NC(Cl)=CC(Cl)=N1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals
Melting point38°C to 43°C
Appearance (Color)White to yellow to orange
GC>=97.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only