alpha-Bromo-o-tolunitrile, 98%
alpha-Bromo-o-tolunitrile, 98%
alpha-Bromo-o-tolunitrile, 98%
Thermo Scientific Chemicals

alpha-Bromo-o-tolunitrile, 98%

CAS: 22115-41-9 | C8H6BrN | 196.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Catalog number 163130250
Price (EUR)
109,00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (EUR)
109,00
Each
Chemical Identifiers
CAS22115-41-9
IUPAC Name2-(bromomethyl)benzonitrile
Molecular FormulaC8H6BrN
InChI KeyQGXNHCXKWFNKCG-UHFFFAOYSA-N
SMILESBrCC1=CC=CC=C1C#N
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to grey-brown
Infrared spectrumConforms
GC>=97.5 %
Appearance (Form)Crystalline powder and/or chunks
Melting point70°C to 74°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only