2,5-Dichlorobenzoic acid, 97%
2,5-Dichlorobenzoic acid, 97%
2,5-Dichlorobenzoic acid, 97%
Thermo Scientific Chemicals

2,5-Dichlorobenzoic acid, 97%

CAS: 50-79-3 | C7H4Cl2O2 | 191.01 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
50 g
250 g
Packaging:
View all
Glass Bottle
Catalog number 163240100
Price (USD)
-
Quantity:
10 g
Packaging:
Glass Bottle
Request bulk or custom format
Chemical Identifiers
CAS50-79-3
IUPAC Name2,5-dichlorobenzoic acid
Molecular FormulaC7H4Cl2O2
InChI KeyQVTQYSFCFOGITD-UHFFFAOYSA-N
SMILESOC(=O)C1=CC(Cl)=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point152°C to 157°C
Appearance (Form)Powder
Titration with NaOH>=96.0 %
Appearance (Color)White to beige to light brown
Infrared spectrumConforms
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only