2-Chlorobenzyl alcohol, 99%
2-Chlorobenzyl alcohol, 99%
2-Chlorobenzyl alcohol, 99%
Thermo Scientific Chemicals

2-Chlorobenzyl alcohol, 99%

CAS: 17849-38-6 | C7H7ClO | 142.58 g/mol
Quantity:
10 g
50 g
Catalog number 163420500
Price (JPY)
-
Quantity:
50 g
Request bulk or custom format
Chemical Identifiers
CAS17849-38-6
IUPAC Name(2-chlorophenyl)methanol
Molecular FormulaC7H7ClO
InChI KeyMBYQPPXEXWRMQC-UHFFFAOYSA-N
SMILESOCC1=CC=CC=C1Cl
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Melting point69°C to 72°C
Appearance (Color)White
GC>=98.5 %
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only